siRNA Details
| Gene Symbol: CCDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282658.1 | NP_001269587.1 | 7 | 1605 | ||
| NM_031455.3 | NP_113643.1 | 3 | 1040 | |||
| GenBank mRNA | AK024578.1 | - | 1 | 23 | ||
| AK095792.1 | - | 7 | 1484 | |||
| AK123393.1 | - | 1 | 1181 | |||
| AK124195.1 | - | 1 | 743 | |||
| AK307203.1 | - | 3 | 832 | |||
| AK307304.1 | - | 3 | 670 | |||
| AL136562.1 | - | 3 | 956 | |||
| BC051334.1 | - | 3 | 1040 | 6302570 | ||

CCDC3
Genomic Map