siRNA Details
| Gene Symbol: KRTAP4-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031854.2 | NP_114060.1 | 1 | 926 | ||
| GenBank mRNA | AJ406943.1 | - | 1 | 926 | ||
| BC004180.2 | - | 1 | 906 | 2959821 | ||
| BC004212.2 | - | 1 | 906 | 2959821 | ||
| BC101159.1 | - | 1 | 918 | 40015747 | ||
| BC101160.1 | - | 1 | 918 | 40015748 | ||
| BC101161.1 | - | 2 | 453 | |||
| BC101162.1 | - | 1 | 918 | 40015750 | ||

KRTAP4-12
Genomic Map