siRNA Details
| Gene Symbol: C1QTNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031908.4 | NP_114114.2 | 3 | 1409 | ||
| XM_011534425.2 | XP_011532727.1 | 4 | 1541 | |||
| XM_011534427.2 | XP_011532729.1 | 5 | 1432 | |||
| GenBank mRNA | BC011699.2 | - | 3 | 1390 | ||

C1QTNF2
Genomic Map