siRNA Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031925.2 | NP_114131.1 | 11, 12 | 1021 | ||
| GenBank mRNA | AF327923.1 | - | 11, 12 | 982 | ||
| BC051850.1 | - | 11, 12 | 1006 | 6201334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM120A| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031925.2 | NP_114131.1 | 11, 12 | 1021 | ||
| GenBank mRNA | AF327923.1 | - | 11, 12 | 982 | ||
| BC051850.1 | - | 11, 12 | 1006 | 6201334 | ||