siRNA Details
| Gene Symbol: CDCA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031942.4 | NP_114148.3 | 5, 6 | 827 | ||
| NM_145810.2 | NP_665809.1 | 4, 5 | 590 | |||
| XM_011511957.2 | XP_011510259.1 | 5, 6 | 1368 | |||
| GenBank mRNA | AK027628.1 | - | 5, 6 | 799 | ||
| AK027642.1 | - | 4, 5 | 571 | |||
| AK223524.1 | - | 4, 5 | 595 | |||
| AK297097.1 | - | 3, 4 | 425 | |||
| AK297245.1 | - | 5, 6 | 687 | |||
| AK300949.1 | - | 5, 6 | 656 | |||
| AY029179.1 | - | 4, 5 | 603 | |||
| BC027966.1 | - | 4, 5 | 556 | 5227112 | ||

CDCA7
Genomic Map