siRNA Details
| Gene Symbol: WDR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291088.1 | NP_001278017.1 | 4 | 955 | ||
| NM_031951.4 | NP_114157.4 | 4 | 838 | |||
| XM_011527360.2 | XP_011525662.1 | 4 | 782 | |||
| GenBank mRNA | AK128826.1 | - | 4 | 809 | ||
| AK302913.1 | - | 4 | 847 | |||
| AY026504.2 | - | 4 | 867 | |||

WDR87
Genomic Map