siRNA Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170722.1 | NP_001164193.1 | 3 | 265 | ||
| NM_001170723.1 | NP_001164194.1 | 3 | 241 | |||
| NM_001170724.1 | NP_001164195.1 | 3 | 241 | |||
| NM_032126.4 | NP_115502.2 | 3 | 241 | |||
| GenBank mRNA | AK093269.1 | - | 3 | 188 | ||
| AL136694.1 | - | 3 | 245 | |||
| AL552172.3 | - | 3 | 163 | |||
| BC034972.2 | - | 3 | 198 | 4823687 | ||
| BI461686.1 | - | 3 | 206 | |||
| BI825610.1 | - | 3 | 175 | |||

TEX35
Genomic Map