siRNA Details
| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032130.2 | NP_115506.1 | 1, 2 | 277 | ||
| XM_006719625.2 | XP_006719688.1 | 1, 2 | 266 | |||
| XM_006719626.2 | XP_006719689.1 | 1, 2 | 266 | |||
| XM_006719627.3 | XP_006719690.1 | 1, 2 | 266 | |||
| XM_011538796.2 | XP_011537098.1 | 1, 2 | 266 | |||
| XM_011538797.2 | XP_011537099.1 | 1, 2 | 266 | |||
| GenBank mRNA | AK093242.1 | - | 1, 2 | 266 | ||
| AL136748.1 | - | 1, 2 | 198 | |||
| BC035621.1 | - | 1, 2 | 235 | 5171752 | ||
| DB087952.1 | - | 1, 2 | 279 | |||

FAM186B
Genomic Map