siRNA Details
| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032048.2 | NP_114437.2 | 4 | 698 | ||
| XM_017026038.1 | XP_016881527.1 | 4 | 685 | |||
| GenBank mRNA | AF270513.1 | - | 4 | 698 | ||
| AL552384.3 | - | 4 | 468 | |||
| BC136541.1 | - | 4 | 652 | 9020531 | ||

EMILIN2
Genomic Map