siRNA Details
| Gene Symbol: RNF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032015.4 | NP_114404.1 | 1 | 2103 | ||
| GenBank mRNA | AK075379.1 | - | 1 | 1861 | ||
| AK122642.1 | - | 1 | 2076 | |||
| BC000058.2 | - | 1 | 1872 | 3507662 | ||
| BC007534.1 | - | 1 | 1843 | 3028875 | ||
| BX647424.1 | - | 1 | 2076 | |||

RNF26
Genomic Map