siRNA Details
| Gene Symbol: IFI27L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032036.2 | NP_114425.1 | 4 | 451 | ||
| GenBank mRNA | AF208232.1 | - | 4 | 417 | ||
| AF238978.1 | - | 2 | 284 | |||
| BC022800.2 | - | 2 | 359 | |||
| BC032626.1 | - | 4 | 410 | 5229498 | ||

IFI27L2
Genomic Map