siRNA Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032120.3 | NP_115496.2 | 4 | 547 | ||
| XM_005250636.4 | XP_005250693.1 | 4 | 611 | |||
| XM_006716149.3 | XP_006716212.1 | 4 | 562 | |||
| XM_006716150.3 | XP_006716213.1 | 4 | 562 | |||
| XM_017012709.1 | XP_016868198.1 | 3 | 559 | |||
| GenBank mRNA | AK294626.1 | - | 4 | 685 | ||
| AK294812.1 | - | 4 | 519 | |||
| AL136619.1 | - | 4 | 529 | |||
| BC033561.1 | - | 4 | 526 | |||
| BC044570.1 | - | 4 | 530 | 4793676 | ||
| BC065716.1 | - | 4 | 517 | |||
| BC104865.1 | - | 4 | 547 | 8143868 | ||
| BC104867.1 | - | 4 | 547 | 8143870 | ||
| BC143576.1 | - | 4 | 548 | 9052087 | ||
| CR533549.1 | - | 4 | 506 | |||
| CR933641.1 | - | 4 | 535 | |||
| CR936726.1 | - | 4 | 526 | |||

RBM48
Genomic Map