siRNA Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF251699.1 | - | 6 | 847 | ||
| AK222642.1 | - | 6 | 831 | |||
| AL136683.1 | - | 6 | 866 | |||
| BC011579.1 | - | 6 | 849 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM222| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF251699.1 | - | 6 | 847 | ||
| AK222642.1 | - | 6 | 831 | |||
| AL136683.1 | - | 6 | 866 | |||
| BC011579.1 | - | 6 | 849 | |||