siRNA Details
| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098794.1 | NP_001092264.1 | 12 | 3342 | ||
| NM_032127.3 | NP_115503.2 | 12 | 3384 | |||
| XM_006718343.2 | XP_006718406.1 | 12 | 3306 | |||
| XM_011520397.2 | XP_011518699.1 | 12 | 3320 | |||
| GenBank mRNA | AB051546.1 | - | 12 | 3247 | ||
| AL136723.1 | - | 12 | 3318 | |||
| BC030825.1 | - | 12 | 3255 | 4811942 | ||

FAM160A2
Genomic Map