siRNA Details
| Gene Symbol: QRICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032134.2 | NP_115510.1 | 4 | 3072 | ||
| XM_005257728.3 | XP_005257785.1 | 4 | 4082 | |||
| XM_006722136.3 | XP_006722199.1 | 1 | 271 | |||
| XM_011525344.2 | XP_011523646.1 | 2 | 2755 | |||
| XM_017025206.1 | XP_016880695.1 | 4 | 4082 | |||
| XM_017025207.1 | XP_016880696.1 | 3 | 4022 | |||
| GenBank mRNA | AL136774.1 | - | 4 | 3072 | ||

QRICH2
Genomic Map