siRNA Details
| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091344.1 | - | 1 | 2365 | ||
| AK299614.1 | - | 3 | 1999 | |||
| AL136782.1 | - | 1 | 2277 | |||
| BC022033.2 | - | 1 | 2360 | 4825536 | ||
| FJ150424.1 | - | 1 | 2360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KBTBD7| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091344.1 | - | 1 | 2365 | ||
| AK299614.1 | - | 3 | 1999 | |||
| AL136782.1 | - | 1 | 2277 | |||
| BC022033.2 | - | 1 | 2360 | 4825536 | ||
| FJ150424.1 | - | 1 | 2360 | |||