siRNA Details
| Gene Symbol: ENKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032140.1 | NP_115516.1 | 3 | 576 | ||
| GenBank mRNA | AL136786.1 | - | 3 | 576 | ||
| BC008284.1 | - | 3 | 543 | 3638194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENKD1| Gene Symbol: ENKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032140.1 | NP_115516.1 | 3 | 576 | ||
| GenBank mRNA | AL136786.1 | - | 3 | 576 | ||
| BC008284.1 | - | 3 | 543 | 3638194 | ||