siRNA Details
| Gene Symbol: DSEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032160.2 | NP_115536.1 | 2 | 1331 | ||
| GenBank mRNA | AF480435.1 | - | 2 | 1583 | ||
| AK021849.1 | - | 1 | 741 | |||
| BC062557.1 | - | 1 | 415 | |||
| BC117325.1 | - | 1 | 145 | 40125876 | ||

DSEL
Genomic Map