siRNA Details
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015574.1 | NP_056389.1 | 1 | 513 | ||
NM_032217.4 | NP_115593.3 | 1 | 513 | |||
NM_198889.2 | NP_942592.1 | 1 | 513 | |||
XM_005265671.3 | XP_005265728.1 | 1 | 857 | |||
XM_005265672.3 | XP_005265729.1 | 1 | 857 | |||
XM_005265673.3 | XP_005265730.1 | 1 | 857 | |||
XM_017008014.1 | XP_016863503.1 | 1 | 857 | |||
XM_017008016.1 | XP_016863505.1 | 1 | 857 | |||
GenBank mRNA | AB014597.2 | - | 1 | 37 | ||
AF308285.1 | - | 1 | 500 | |||
AK307581.1 | - | 1 | 513 | |||
BC004173.2 | - | 1 | 494 | |||
BC043394.1 | - | 1 | 482 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198010, NM_030886, AK031350, BC085166.
Panther Classification:
Gene Ontology Categories:
