siRNA Details
| Gene Symbol: ELAVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001420.3 | NP_001411.2 | 7 | 2538 | ||
| NM_032281.2 | NP_115657.2 | 7 | 2517 | |||
| GenBank mRNA | BC011875.2 | - | 1 | 285 | 3161837 | |
| BC080533.1 | - | 1 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELAVL3| Gene Symbol: ELAVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001420.3 | NP_001411.2 | 7 | 2538 | ||
| NM_032281.2 | NP_115657.2 | 7 | 2517 | |||
| GenBank mRNA | BC011875.2 | - | 1 | 285 | 3161837 | |
| BC080533.1 | - | 1 | 283 | |||