siRNA Details
Gene Symbol: PRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039091.2 | NP_001034180.1 | 7 | 1420 | ||
NM_002765.4 | NP_002756.1 | 7 | 1411 | |||
GenBank mRNA | AK310332.1 | - | 5 | 833 | ||
BC030019.2 | - | 7 | 1352 | |||
BC040483.1 | - | 7 | 1354 | 5272983 | ||
BC110875.1 | - | 7 | 1345 | 6472836 | ||
Y00971.1 | - | 7 | 1328 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
