siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287498.1 | NP_001274427.1 | 7 | 739 | ||
| NM_032295.3 | NP_115671.1 | 7 | 739 | |||
| NM_207113.2 | NP_996996.1 | 7 | 739 | |||
| XM_011516626.2 | XP_011514928.1 | 9 | 1236 | |||
| XM_011516627.2 | XP_011514929.1 | 9 | 1235 | |||
| XM_017012712.1 | XP_016868201.1 | 9 | 1212 | |||
| XM_017012713.1 | XP_016868202.1 | 9 | 1231 | |||
| XM_017012715.1 | XP_016868204.1 | 9 | 1255 | |||
| XM_017012716.1 | XP_016868205.1 | 9 | 1259 | |||
| XM_017012717.1 | XP_016868206.1 | 9 | 1231 | |||
| XM_017012718.1 | XP_016868207.1 | 9 | 1230 | |||
| GenBank mRNA | AK074823.1 | - | 7 | 700 | ||
| AK296543.1 | - | 6 | 609 | |||
| AK299276.1 | - | 7 | 683 | |||
| AK300265.1 | - | 6 | 551 | |||
| AL136583.1 | - | 7 | 646 | |||
| BC028380.1 | - | 7 | 689 | 4811814 | ||
| BC046567.1 | - | 7 | 691 | 5246993 | ||

SLC37A3
Genomic Map