siRNA Details
| Gene Symbol: ZSCAN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199479.1 | NP_001186408.1 | 2 | 305 | ||
| NM_001199480.1 | NP_001186409.1 | 2 | 269 | |||
| NM_006299.4 | NP_006290.1 | 2 | 305 | |||
| XM_011514875.2 | XP_011513177.1 | 2 | 782 | |||
| XM_011514876.2 | XP_011513178.1 | 2 | 781 | |||
| XM_011514878.2 | XP_011513180.1 | 2 | 777 | |||
| XM_017011267.1 | XP_016866756.1 | 2 | 756 | |||
| GenBank mRNA | AK226035.1 | - | 2 | 261 | ||
| AK304013.1 | - | 2 | 281 | |||
| AU310237.1 | - | 2 | 282 | |||
| AY261373.1 | - | 2 | 217 | |||
| AY550973.1 | - | 1 | 183 | |||
| BC010374.1 | - | 2 | 265 | 4132536 | ||
| U62392.1 | - | 2 | 217 | |||

ZSCAN9
Genomic Map