siRNA Details
| Gene Symbol: PSMG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134340.1 | NP_001127812.1 | 2, 3 | 263 | ||
| NM_032302.3 | NP_115678.1 | 1, 2 | 872 | |||
| GenBank mRNA | BC004308.1 | - | 1, 2 | 451 | 3627097 | |
| BC027171.1 | - | 1, 2 | 873 | 4344108 | ||
| BM825255.1 | - | 2, 3 | 276 | |||
| CN261807.1 | - | 1, 2 | 257 | |||

PSMG3
Genomic Map