siRNA Details
| Gene Symbol: FBXO30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032145.4 | NP_115521.3 | 2 | 1471 | ||
| XM_005267159.3 | XP_005267216.1 | 2 | 1310 | |||
| XM_011536177.2 | XP_011534479.1 | 2 | 6330 | |||
| XM_017011353.1 | XP_016866842.1 | 2 | 1310 | |||
| GenBank mRNA | AF248640.1 | - | 1 | 352 | ||
| AK124138.1 | - | 1 | 671 | |||
| AK313613.1 | - | 1 | 262 | |||
| BC024326.1 | - | 2 | 1439 | 3915249 | ||

FBXO30
Genomic Map