siRNA Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032193.3 | NP_115569.2 | 4 | 677 | ||
| GenBank mRNA | AF346606.1 | - | 4 | 677 | ||
| AK024627.1 | - | 4 | 534 | |||
| AK225131.1 | - | 4 | 534 | |||
| AK297660.1 | - | 4 | 447 | |||
| BC023588.2 | - | 4 | 557 | 4763204 | ||
| BM921525.1 | - | 4 | 546 | |||
| L08647.1 | - | 3 | 406 | |||

RNASEH2C
Genomic Map