siRNA Details
| Gene Symbol: C19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288834.1 | NP_001275763.1 | 2 | 671 | ||
| NM_032207.3 | NP_115583.1 | 2 | 671 | |||
| XM_006722920.3 | XP_006722983.1 | 2 | 671 | |||
| XM_011528354.2 | XP_011526656.1 | 2 | 671 | |||
| XM_017027350.1 | XP_016882839.1 | 2 | 671 | |||
| GenBank mRNA | AK025395.1 | - | 2 | 569 | ||
| AK225118.1 | - | 2 | 588 | |||
| AK225678.1 | - | 2 | 581 | |||
| BC027869.1 | - | 2 | 543 | 5205152 | ||

C19orf44
Genomic Map