siRNA Details
| Gene Symbol: TMEM126A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244735.1 | NP_001231664.1 | 2 | 310 | ||
| NM_032273.3 | NP_115649.1 | 3 | 403 | |||
| GenBank mRNA | AK312081.1 | - | 3 | 335 | ||
| AL136941.1 | - | 3 | 349 | |||
| BC007875.2 | - | 3 | 321 | 3948518 | ||
| BQ943238.1 | - | 3 | 347 | |||
| DB454483.1 | - | 3 | 403 | |||

TMEM126A
Genomic Map