siRNA Details
| Gene Symbol: TCHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143852.1 | NP_001137324.1 | 3 | 380 | ||
| NM_032300.4 | NP_115676.1 | 3 | 441 | |||
| XM_011538836.2 | XP_011537138.1 | 3 | 716 | |||
| XM_011538837.2 | XP_011537139.1 | 3 | 289 | |||
| XM_017020024.1 | XP_016875513.1 | 3 | 380 | |||
| XM_017020025.1 | XP_016875514.1 | 3 | 404 | |||
| GenBank mRNA | AK092736.1 | - | 3 | 380 | ||
| AK301262.1 | - | 3 | 358 | |||
| AY007230.1 | - | 3 | 442 | |||
| BC004285.1 | - | 3 | 361 | 3617849 | ||
| BC022838.1 | - | 3 | 670 | |||

TCHP
Genomic Map