siRNA Details
| Gene Symbol: C4orf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032149.2 | NP_115525.2 | 4, 5 | 772 | ||
| XM_011532315.1 | XP_011530617.1 | 4, 5 | 752 | |||
| GenBank mRNA | AK093085.1 | - | 4, 5 | 764 | ||
| AK223345.1 | - | 4, 5 | 759 | |||
| AK314711.1 | - | 4, 5 | 755 | |||
| AL136838.1 | - | 4, 5 | 733 | |||
| BC074759.2 | - | 3, 4 | 449 | 30915218 | ||
| CR533542.1 | - | 3, 4 | 410 | |||

C4orf17
Genomic Map