siRNA Details
| Gene Symbol: ZIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168378.1 | NP_001161850.1 | 5 | 2228 | ||
| NM_001168379.1 | NP_001161851.1 | 5 | 2055 | |||
| NM_001243256.1 | NP_001230185.1 | 4 | 1736 | |||
| NM_032153.5 | NP_115529.2 | 5 | 2354 | |||
| NR_033118.2 | 3 | 1665 | ||||
| NR_033119.2 | 3 | 1611 | ||||
| NR_040762.1 | 3 | 1512 | ||||
| GenBank mRNA | AK056262.1 | - | 5 | 2165 | ||
| AK056394.1 | - | 3 | 2539 | |||
| AK094286.1 | - | 3 | 1851 | |||
| AK124600.1 | - | 1 | 2690 | |||
| AK126595.1 | - | 2 | 2891 | |||
| BC126267.1 | - | 5 | 1990 | 8991983 | ||
| BC136339.1 | - | 5 | 1990 | 9020326 | ||

ZIC4
Genomic Map