siRNA Details
| Gene Symbol: MFSD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294341.1 | NP_001281270.1 | 9 | 1486 | ||
| NM_001294342.1 | NP_001281271.1 | 7 | 1129 | |||
| NM_032219.3 | NP_115595.2 | 9 | 1483 | |||
| XM_005272297.1 | XP_005272354.1 | 7 | 1219 | |||
| XM_005272299.4 | XP_005272356.1 | 9 | 1513 | |||
| XM_006713918.1 | XP_006713981.1 | 9 | 1140 | |||
| XM_006713919.1 | XP_006713982.1 | 7 | 1222 | |||
| XM_011513568.1 | XP_011511870.1 | 9 | 2290 | |||
| XM_011513571.1 | XP_011511873.1 | 10 | 1310 | |||
| XM_011513572.1 | XP_011511874.1 | 7 | 1306 | |||
| XM_011513573.2 | XP_011511875.1 | 9 | 1513 | |||
| XM_011513574.1 | XP_011511876.1 | 8 | 1279 | |||
| XM_011513575.2 | XP_011511877.1 | 8 | 2058 | |||
| XM_017008700.1 | XP_016864189.1 | 8 | 1393 | |||
| GenBank mRNA | AK025922.1 | - | 7 | 961 | ||
| AK292010.1 | - | 9 | 1381 | |||
| AY203936.1 | - | 5 | 832 | |||
| AY358401.1 | - | 9 | 1188 | |||
| BC030246.1 | - | 9 | 1226 | 5216645 | ||

MFSD7
Genomic Map