siRNA Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 7 | 3653 | ||
NM_032281.2 | NP_115657.2 | 7 | 3632 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001420.3 | NP_001411.2 | 7 | 3653 | ||
NM_032281.2 | NP_115657.2 | 7 | 3632 |