siRNA Details
| Gene Symbol: TCHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143852.1 | NP_001137324.1 | 13 | 2049 | ||
| NM_032300.4 | NP_115676.1 | 13 | 2110 | |||
| XM_011538836.2 | XP_011537138.1 | 13 | 2385 | |||
| XM_011538837.2 | XP_011537139.1 | 13 | 1958 | |||
| XM_017020025.1 | XP_016875514.1 | 13 | 2294 | |||
| GenBank mRNA | AY007230.1 | - | 13 | 2111 | ||
| BC004285.1 | - | 13 | 2030 | 3617849 | ||
| BX648596.1 | - | 2 | 1773 | |||

TCHP
Genomic Map