siRNA Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318010.1 | NP_001304939.1 | 2 | 320 | ||
| NM_001318011.1 | NP_001304940.1 | 2 | 320 | |||
| NM_032310.4 | NP_115686.3 | 2 | 320 | |||
| NR_134464.1 | 2 | 320 | ||||
| NR_134465.1 | 2 | 320 | ||||
| NR_134466.1 | 2 | 320 | ||||
| NR_134467.1 | 2 | 320 | ||||
| NR_134468.1 | 2 | 320 | ||||
| GenBank mRNA | AK057716.1 | - | 2 | 226 | ||
| AK091611.1 | - | 2 | 226 | |||
| BC004500.2 | - | 3 | 189 | 3833318 | ||
| BC038856.1 | - | 2 | 213 | 5215640 | ||
| BC091476.1 | - | 2 | 163 | |||
| BM930683.1 | - | 2 | 165 | |||
| BX354102.2 | - | 2 | 198 | |||
| CD676329.1 | - | 2 | 176 | |||
| DN998463.1 | - | 2 | 213 | |||
| HY005780.1 | - | 2 | 321 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026738, BC147074, BC147073, AK154517, AK003526, and others.

CARD19
Genomic Map