siRNA Details
| Gene Symbol: WDR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099737.2 | NP_001093207.1 | 5, 6 | 686 | ||
| NM_032332.3 | NP_115708.1 | 3, 4 | 429 | |||
| NR_029375.1 | 5, 6 | 669 | ||||
| GenBank mRNA | AK074525.1 | - | 5, 6 | 659 | ||
| AK223195.1 | - | 3, 4 | 379 | |||
| AK314164.1 | - | 5, 6 | 663 | |||
| BC005870.2 | - | 3, 4 | 362 | 2961616 | ||
| CB143206.1 | - | 3, 4 | 429 | |||

WDR83
Genomic Map