siRNA Details
| Gene Symbol: LLPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032338.3 | NP_115714.1 | 2 | 136 | ||
| GenBank mRNA | AK057947.1 | - | 1 | 49 | ||
| AK091568.1 | - | 2 | 136 | |||
| BC006002.1 | - | 2 | 132 | 4247279 | ||
| BC070232.1 | - | 2 | 128 | 4594274 | ||
| BC107780.1 | - | 2 | 137 | 6698895 | ||

LLPH
Genomic Map