siRNA Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032377.3 | NP_115753.1 | 4 | 925 | ||
| XM_017027356.1 | XP_016882845.1 | 5 | 1080 | |||
| XM_017027357.1 | XP_016882846.1 | 4 | 939 | |||
| XM_017027358.1 | XP_016882847.1 | 4 | 1079 | |||
| XM_017027359.1 | XP_016882848.1 | 4 | 995 | |||
| GenBank mRNA | AK001171.1 | - | 5 | 1076 | ||
| BC007516.2 | - | 3 | 874 | 3028469 | ||
| BG424379.1 | - | 406 | ||||

ELOF1
Genomic Map