siRNA Details
| Gene Symbol: C15orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032413.3 | NP_115789.1 | 4 | 515 | ||
| NM_197955.2 | NP_922946.1 | 5 | 401 | |||
| GenBank mRNA | AB026707.1 | - | 5 | 295 | ||
| AF228422.1 | - | 4 | 436 | |||
| AK026298.1 | - | 4 | 433 | |||
| BC021173.2 | - | 5 | 321 | 5269504 | ||
| BM312626.1 | - | 5 | 365 | |||
| CA865860.1 | - | 5 | 407 | |||
| DB447060.1 | - | 5 | 403 | |||

C15orf48
Genomic Map