siRNA Details
| Gene Symbol: CHCHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304353.1 | NP_001291282.1 | 4 | 407 | ||
| NM_001304354.1 | NP_001291283.1 | 4 | 404 | |||
| NM_032309.3 | NP_115685.1 | 4 | 570 | |||
| GenBank mRNA | AK096990.1 | - | 3 | 2913 | ||
| BC004498.2 | - | 4 | 548 | 3831856 | ||
| BM563093.1 | - | 4 | 404 | |||
| BX115079.1 | - | 4 | 406 | |||
| N39730.1 | - | 4 | 404 | |||

CHCHD5
Genomic Map