siRNA Details
| Gene Symbol: DYDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270041.1 | NP_001256970.1 | 3 | 249 | ||
| NM_001270042.1 | NP_001256971.1 | 3 | 262 | |||
| NM_032372.5 | NP_115748.1 | 4 | 402 | |||
| NR_070308.1 | 5 | 780 | ||||
| NR_070309.1 | 4 | 562 | ||||
| XM_011540267.2 | XP_011538569.1 | 3 | 221 | |||
| XM_011540268.1 | XP_011538570.1 | 3 | 211 | |||
| XM_011540270.2 | XP_011538572.1 | 3 | 222 | |||
| GenBank mRNA | AK093378.1 | - | 4 | 370 | ||
| BC007374.2 | - | 4 | 548 | 3637472 | ||
| BC018606.2 | - | 5 | 780 | |||
| BQ229460.1 | - | 3 | 273 | |||
| DA590784.1 | - | 3 | 249 | |||

DYDC2
Genomic Map