siRNA Details
| Gene Symbol: PHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015877.1 | NP_001015877.1 | 3 | 426 | ||
| NM_032335.3 | NP_115711.2 | 3 | 426 | |||
| NM_032458.2 | NP_115834.1 | 3 | 426 | |||
| GenBank mRNA | AB058726.1 | - | 3 | 312 | ||
| AK290095.1 | - | 3 | 397 | |||
| AL832725.1 | - | 3 | 419 | |||
| AY157622.1 | - | 3 | 426 | |||
| BC005994.1 | - | 3 | 365 | 4064169 | ||
| CD685336.1 | - | 3 | 434 | |||
| DR000282.1 | - | 3 | 461 | |||

PHF6
Genomic Map