siRNA Details
| Gene Symbol: COQ5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032314.3 | NP_115690.3 | 5, 6 | 798 | ||
| XM_006719639.2 | XP_006719702.1 | 6, 7 | 898 | |||
| GenBank mRNA | AK222610.1 | - | 5, 6 | 788 | ||
| AK225242.1 | - | 5, 6 | 788 | |||
| AK293656.1 | - | 5, 6 | 819 | |||
| AK298214.1 | - | 4, 5 | 576 | |||
| AK315681.1 | - | 5, 6 | 789 | |||
| BC004916.2 | - | 5, 6 | 774 | |||
| BC107874.1 | - | 5, 6 | 792 | 6452052 | ||
| BX647562.1 | - | 5, 6 | 787 | |||
| EU700459.1 | - | 5, 6 | 792 | |||

COQ5
Genomic Map