siRNA Details
| Gene Symbol: UTP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032334.2 | NP_115710.2 | 3 | 655 | ||
| GenBank mRNA | AK315770.1 | - | 3 | 599 | ||
| BC005955.1 | - | 3 | 655 | 4285502 | ||
| BC022441.1 | - | 3 | 655 | 4285036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UTP23| Gene Symbol: UTP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032334.2 | NP_115710.2 | 3 | 655 | ||
| GenBank mRNA | AK315770.1 | - | 3 | 599 | ||
| BC005955.1 | - | 3 | 655 | 4285502 | ||
| BC022441.1 | - | 3 | 655 | 4285036 | ||