siRNA Details
| Gene Symbol: C7orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134395.1 | NP_001127867.1 | 5 | 977 | ||
| NM_001134396.1 | NP_001127868.1 | 5 | 958 | |||
| NM_001318252.1 | NP_001305181.1 | 5 | 1861 | |||
| NM_032350.5 | NP_115726.1 | 5 | 987 | |||
| NR_134537.1 | 4 | 963 | ||||
| XM_006715793.2 | XP_006715856.1 | 5 | 1389 | |||
| XM_017012722.1 | XP_016868211.1 | 6 | 808 | |||
| GenBank mRNA | AF495721.1 | - | 5 | 1817 | ||
| BC006224.2 | - | 5 | 987 | 3941780 | ||

C7orf50
Genomic Map