siRNA Details
| Gene Symbol: CCDC115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321118.1 | NP_001308047.1 | 5 | 1160 | ||
| NM_001321119.1 | NP_001308048.1 | 5 | 682 | |||
| NM_032357.3 | NP_115733.2 | 5 | 981 | |||
| NR_104471.1 | 6 | 1363 | ||||
| NR_135548.1 | 6 | 1184 | ||||
| GenBank mRNA | AK054693.1 | - | 5 | 947 | ||
| AK092196.1 | - | 1 | 1684 | |||
| AK124405.1 | - | 2 | 2869 | |||
| AK295922.1 | - | 6 | 1020 | |||
| BC006429.1 | - | 5 | 743 | 3350642 | ||

CCDC115
Genomic Map