siRNA Details
| Gene Symbol: PROK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032414.2 | NP_115790.1 | 3 | 1174 | ||
| GenBank mRNA | AF333024.1 | - | 3 | 1176 | ||
| AY029225.1 | - | 3 | 1171 | |||
| AY358683.1 | - | 3 | 1213 | |||
| BC025399.1 | - | 3 | 1151 | 4826496 | ||

PROK1
Genomic Map