siRNA Details
| Gene Symbol: PHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015877.1 | NP_001015877.1 | 6 | 703 | ||
| NM_032335.3 | NP_115711.2 | 6 | 706 | |||
| NM_032458.2 | NP_115834.1 | 6 | 703 | |||
| GenBank mRNA | AB058726.1 | - | 6 | 589 | ||
| AK290095.1 | - | 6 | 674 | |||
| AK303369.1 | - | 5 | 566 | |||
| AL832725.1 | - | 6 | 699 | |||
| AY157622.1 | - | 6 | 703 | |||
| BC005994.1 | - | 6 | 645 | 4064169 | ||

PHF6
Genomic Map