siRNA Details
| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032483.3 | NP_115872.2 | 6, 7 | 663 | ||
| GenBank mRNA | AF212238.1 | - | 6, 7 | 617 | ||
| BC106014.1 | - | 6, 7 | 633 | 3877263 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLPP5| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032483.3 | NP_115872.2 | 6, 7 | 663 | ||
| GenBank mRNA | AF212238.1 | - | 6, 7 | 617 | ||
| BC106014.1 | - | 6, 7 | 633 | 3877263 | ||