siRNA Details
| Gene Symbol: ST6GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142351.1 | NP_001135823.1 | 2 | 508 | ||
| NM_001142352.1 | NP_001135824.1 | 2 | 578 | |||
| NM_001322362.1 | NP_001309291.1 | 2 | 557 | |||
| NM_032528.2 | NP_115917.1 | 2 | 578 | |||
| XM_006712802.1 | XP_006712865.1 | 2 | 593 | |||
| XM_011511999.2 | XP_011510301.1 | 2 | 690 | |||
| XM_011512000.2 | XP_011510302.1 | 2 | 775 | |||
| XM_011512001.2 | XP_011510303.1 | 2 | 775 | |||
| XM_017005110.1 | XP_016860599.1 | 2 | 529 | |||
| GenBank mRNA | AB058780.1 | - | 2 | 508 | ||
| AB059555.1 | - | 2 | 564 | |||
| AK307784.1 | - | 2 | 579 | |||
| BC008680.1 | - | 2 | 564 | 3856171 | ||

ST6GAL2
Genomic Map